Assay Details
Gene Symbol: Stard6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007627.1 | NP_001007628.1 | 5 - 6 | 494 | 125 | |
XM_006254921.2 | XP_006254983.1 | 6 - 7 | 716 | 125 | ||
XM_008772111.1 | XP_008770333.1 | 6 - 7 | 564 | 125 | ||
XM_008772112.1 | XP_008770334.1 | 5 - 6 | 519 | 125 | ||
GenBank mRNA | AY555189.1 | - | 6 - 7 | 664 | 125 | |
BC078976.1 | - | 5 - 6 | 494 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
