Assay Details
Gene Symbol: Stard6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007627.1 | NP_001007628.1 | 6 - 7 | 615 | 95 | |
XM_006254921.2 | XP_006254983.1 | 7 - 8 | 837 | 95 | ||
XM_008772111.1 | XP_008770333.1 | 7 - 8 | 685 | 95 | ||
XM_008772112.1 | XP_008770334.1 | 6 - 7 | 640 | 95 | ||
GenBank mRNA | BC078976.1 | - | 6 - 7 | 615 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
