Assay Details
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145366.1 | NP_001138838.1 | 2 - 3 | 193 | 87 | |
NM_001145367.1 | NP_001138839.1 | 2 - 3 | 371 | 87 | ||
XM_006237009.2 | XP_006237071.1 | 2 - 3 | 164 | 87 | ||
GenBank mRNA | AF156665.1 | - | 2 - 3 | 193 | 87 | |
AF246457.1 | - | 2 - 3 | 121 | 87 | ||
AF246458.1 | - | 2 - 3 | 161 | 87 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF156665.1, AF246457.1, AF246458.1, NM_001145366.1, NM_001145367.1, XM_006237009.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
