Assay Details
Gene Symbol: Tnfrsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013091.1 | NP_037223.1 | 3 - 4 | 560 | 86 | |
XM_006237334.2 | XP_006237396.1 | 2 - 3 | 795 | 86 | ||
GenBank mRNA | AF329976.1 | - | 3 - 4 | 559 | 86 | |
AF329977.1 | - | 3 - 4 | 559 | 86 | ||
AF329978.1 | - | 3 - 4 | 559 | 86 | ||
AF329979.1 | - | 3 - 4 | 559 | 86 | ||
AF329980.1 | - | 3 - 4 | 559 | 86 | ||
AF329981.1 | - | 3 - 4 | 559 | 86 | ||
BC086413.1 | - | 3 - 4 | 580 | 86 | ||
M63122.1 | - | 3 - 4 | 560 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
