Assay Details
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107890.1 | NP_001101360.1 | 2 - 3 | 622 | 62 | |
XM_006237494.2 | XP_006237556.1 | 6 - 7 | 753 | 62 | ||
XM_006237495.2 | XP_006237557.1 | 5 - 6 | 569 | 62 | ||
XM_006237496.2 | XP_006237558.1 | 5 - 6 | 632 | 62 | ||
XM_006237498.2 | XP_006237560.1 | 4 - 5 | 452 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
