Assay Details
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107890.1 | NP_001101360.1 | 5 - 6 | 934 | 94 | |
XM_006237494.2 | XP_006237556.1 | 9 - 10 | 1065 | 94 | ||
XM_006237495.2 | XP_006237557.1 | 8 - 9 | 881 | 94 | ||
XM_006237496.2 | XP_006237558.1 | 8 - 9 | 944 | 94 | ||
XM_006237498.2 | XP_006237560.1 | 7 - 8 | 764 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
