Assay Details
Gene Symbol: Slc6a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133623.1 | NP_598307.1 | 13 - 14 | 1641 | 89 | |
XM_006237218.2 | XP_006237280.1 | 13 - 14 | 1658 | 89 | ||
XM_006237219.2 | XP_006237281.1 | 14 - 15 | 1774 | 89 | ||
XM_006237220.2 | XP_006237282.1 | 13 - 14 | 1640 | 89 | ||
XM_006237221.2 | XP_006237283.1 | 14 - 15 | 1756 | 89 | ||
XM_006237222.2 | XP_006237284.1 | 12 - 13 | 1512 | 89 | ||
XM_006237223.2 | XP_006237285.1 | 12 - 13 | 1639 | 89 | ||
GenBank mRNA | M95762.1 | - | 13 - 14 | 1641 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
