Assay Details
Gene Symbol: Slc6a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133623.1 | NP_598307.1 | 7 - 8 | 953 | 97 | |
XM_006237218.2 | XP_006237280.1 | 7 - 8 | 955 | 97 | ||
XM_006237219.2 | XP_006237281.1 | 8 - 9 | 1071 | 97 | ||
XM_006237220.2 | XP_006237282.1 | 7 - 8 | 937 | 97 | ||
XM_006237221.2 | XP_006237283.1 | 8 - 9 | 1053 | 97 | ||
XM_006237222.2 | XP_006237284.1 | 6 - 7 | 809 | 97 | ||
XM_006237223.2 | XP_006237285.1 | 6 - 7 | 936 | 97 | ||
GenBank mRNA | M95762.1 | - | 7 - 8 | 953 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
