Assay Details
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763441.1 | XP_008761663.1 | 7 - 8 | 1398 | 95 | |
XM_008763442.1 | XP_008761664.1 | 9 - 10 | 1386 | 95 | ||
XM_008763443.1 | XP_008761665.1 | 10 - 11 | 2141 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
