Assay Details
Gene Symbol: Bcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017253.2 | NP_058949.1 | 10 - 11 | 1288 | 61 | |
XM_008763370.1 | XP_008761592.1 | 10 - 11 | 1301 | 61 | ||
XM_008763371.1 | XP_008761593.1 | 10 - 11 | 1551 | 61 | ||
XM_008763372.1 | XP_008761594.1 | 11 - 12 | 1377 | 61 | ||
XM_008763373.1 | XP_008761595.1 | 10 - 11 | 1480 | 61 | ||
GenBank mRNA | AF165887.1 | - | 8 - 9 | 1045 | 61 | |
AJ278701.1 | - | 10 - 11 | 1282 | 61 | ||
BC087710.1 | - | 10 - 11 | 1288 | 61 | ||
U35774.1 | - | 10 - 11 | 1254 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
branched-chain amino acid biosynthetic process
branched-chain amino acid catabolic process
branched-chain-amino-acid transaminase activity
identical protein binding
L-leucine transaminase activity
L-valine transaminase activity
L-isoleucine transaminase activity
biological_process
branched-chain amino acid biosynthetic process
branched-chain amino acid catabolic process
branched-chain-amino-acid transaminase activity
identical protein binding
L-leucine transaminase activity
L-valine transaminase activity
L-isoleucine transaminase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P025016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87164 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P025016 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P025016 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P025016 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11881 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700517522 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12049 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46114 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13057 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54385 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71270 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42580 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700517522 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10866832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866833 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866830 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10866819 | - | - |
Rat Genome 230 2.0 Array | 1369520_a_at | - | - |
Rat Genome 230 2.0 Array | 1370869_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369520_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1370869_at | - | - |
Affymetrix HT Rat Focus Array | 1370869_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370869_at | - | - |
Affymetrix HT Rat Focus Array | 1369520_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369520_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400028863.rn.1 | TC0400004065.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400028864.rn.1 | TC0400004065.rn.1 | RTA TC |
Rat Expression Set 230 | 1370869_at | - | - |
Rat Expression Set 230 | 1369520_PM_a_at | - | - |
Rat Expression Set 230 | 1370869_PM_at | - | - |
Rat Expression Set 230 | 1369520_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866819 | - | - |
Rat Genome U34 Set | U35774_at | - | - |
Rat Genome U34 Set | rc_AA900881_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22165252 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040300 | - | - |
RatRef-12 Expression BeadChip | ILMN_161861 | - | - |
