Assay Details
Gene Symbol: Ltbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008315.1 | NP_001008316.1 | 3 - 4 | 506 | 157 | |
XM_006237351.1 | XP_006237413.1 | 3 - 4 | 556 | 157 | ||
GenBank mRNA | BC085880.1 | - | 3 - 4 | 506 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
