Assay Details
Gene Symbol: Ldhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012595.1 | NP_036727.1 | 2 - 3 | 155 | 84 | |
XM_006237615.2 | XP_006237677.1 | 3 - 4 | 326 | 84 | ||
GenBank mRNA | BC059149.1 | - | 2 - 3 | 212 | 84 | |
FQ212899.1 | - | 2 - 3 | 218 | 84 | ||
FQ214847.1 | - | 2 - 3 | 202 | 84 | ||
FQ215687.1 | - | 2 - 3 | 218 | 84 | ||
U07181.1 | - | 2 - 3 | 155 | 84 |
Gene Symbol: Hbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FQ213582.1 | - | 200 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
