Assay Details
Gene Symbol: Ldhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC059149.1 | - | 1 - 2 | 80 | 101 | |
FQ212899.1 | - | 1 - 2 | 86 | 101 | ||
FQ214847.1 | - | 1 - 2 | 70 | 101 | ||
FQ215687.1 | - | 1 - 2 | 86 | 101 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC059149.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
