Assay Details
Gene Symbol: Itfg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108648.1 | NP_001102118.1 | 1 - 2 | 209 | 74 | |
XM_006237409.2 | XP_006237471.1 | 1 - 2 | 598 | 74 | ||
XM_006237410.2 | XP_006237472.1 | 1 - 2 | 599 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
