Assay Details
Gene Symbol: Slc25a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044280.2 | NP_001037745.2 | 1 - 2 | 223 | 96 | |
GenBank mRNA | BC094517.1 | - | 1 - 2 | 168 | 96 | |
CB726923.1 | - | 1 - 2 | 224 | 96 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC094517.1, NM_001044280.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
