Assay Details
Gene Symbol: LOC100366044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760951.1 | XP_008759173.1 | 1 | 511 | 129 | |
XM_008760952.1 | XP_008759174.1 | 1 | 511 | 129 |
Gene Symbol: Fxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC083807.1 | - | 514 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
