Assay Details
Gene Symbol: Pde7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031080.1 | NP_112342.1 | 3 - 4 | 416 | 62 | |
XM_006232188.2 | XP_006232250.1 | 3 - 4 | 1107 | 62 | ||
XM_008760880.1 | XP_008759102.1 | 2 - 3 | 1242 | 62 | ||
GenBank mRNA | U77880.1 | - | 2 - 3 | 121 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
