Assay Details
Gene Symbol: Pde7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031080.1 | NP_112342.1 | 8 - 9 | 957 | 109 | |
XM_006232188.2 | XP_006232250.1 | 8 - 9 | 1648 | 109 | ||
XM_008760880.1 | XP_008759102.1 | 7 - 8 | 1783 | 109 | ||
GenBank mRNA | U77880.1 | - | 7 - 8 | 662 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
