Assay Details
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100977.1 | NP_001094447.1 | 3 - 4 | 305 | 76 | |
XM_006232178.2 | XP_006232240.1 | 4 - 5 | 462 | 76 | ||
XM_006232179.2 | XP_006232241.1 | 4 - 5 | 441 | 76 | ||
XM_006232180.2 | XP_006232242.1 | 3 - 4 | 1251 | 76 | ||
GenBank mRNA | EF221637.1 | - | 3 - 4 | 305 | 76 | |
FQ217237.1 | - | 4 - 5 | 413 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
