Assay Details
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100977.1 | NP_001094447.1 | 5 - 6 | 790 | 149 | |
XM_006232178.2 | XP_006232240.1 | 6 - 7 | 941 | 149 | ||
XM_006232179.2 | XP_006232241.1 | 6 - 7 | 920 | 149 | ||
XM_006232180.2 | XP_006232242.1 | 5 - 6 | 1730 | 149 | ||
GenBank mRNA | EF221637.1 | - | 5 - 6 | 790 | 149 | |
FQ217237.1 | - | 6 - 7 | 892 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
