Assay Details
Gene Symbol: Ccna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053702.3 | NP_446154.3 | 7 - 8 | 1307 | 86 | |
XM_006232266.2 | XP_006232328.1 | 8 - 9 | 1444 | 86 | ||
XM_008760903.1 | XP_008759125.1 | 8 - 9 | 1439 | 86 | ||
GenBank mRNA | AY623027.1 | - | 6 - 7 | 1281 | 86 | |
AY623028.1 | - | 7 - 8 | 1686 | 86 | ||
BC127462.1 | - | 8 - 9 | 1400 | 86 | ||
CK845133.1 | - | 1 - 2 | 86 | |||
CV114678.1 | - | 4 - 5 | 574 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
