Assay Details
Gene Symbol: Ercc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107650.2 | NP_001101120.2 | 6 - 7 | 604 | 127 | |
NR_073140.1 | 5 - 6 | 502 | 127 | |||
XM_008760728.1 | XP_008758950.1 | 5 - 6 | 514 | 127 | ||
GenBank mRNA | BC161924.1 | - | 5 - 6 | 479 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
