Assay Details
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254793.2 | XP_006254855.1 | 6 - 7 | 1631 | 109 | |
XM_008772090.1 | XP_008770312.1 | 4 - 5 | 2757 | 109 | ||
GenBank mRNA | AM712909.1 | - | 5 - 6 | 1072 | 109 | |
U20906.1 | - | 5 - 6 | 1092 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
