Assay Details
Gene Symbol: Elf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012181.2 | NP_001012181.1 | 5 - 6 | 1359 | 113 | |
NM_001033909.1 | NP_001029081.1 | 7 - 8 | 1039 | 113 | ||
XM_006232325.2 | XP_006232387.1 | 8 - 9 | 1241 | 113 | ||
XM_006232327.2 | XP_006232389.1 | 5 - 6 | 3427 | 113 | ||
XM_006232328.1 | XP_006232390.1 | 6 - 7 | 753 | 113 | ||
XM_006232329.1 | XP_006232391.1 | 6 - 7 | 717 | 113 | ||
XM_008760992.1 | XP_008759214.1 | 7 - 8 | 1624 | 113 | ||
GenBank mRNA | BC083585.1 | - | 8 - 9 | 1191 | 113 | |
BC083598.1 | - | 5 - 6 | 711 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
