Assay Details
Gene Symbol: Cct3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199091.2 | NP_954522.1 | 10 - 11 | 994 | 93 | |
GenBank mRNA | BC063178.1 | - | 10 - 11 | 998 | 93 | |
FQ213900.1 | - | 3 - 4 | 236 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
