Assay Details
Gene Symbol: Rab13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031092.1 | NP_112354.1 | 3 - 4 | 332 | 136 | |
GenBank mRNA | AF525280.1 | - | 3 - 4 | 332 | 136 | |
FQ222701.1 | - | 3 - 4 | 258 | 136 | ||
M83678.1 | - | 2 - 3 | 131 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
