Assay Details
Gene Symbol: Pdia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105775.2 | NP_001099245.2 | 1427 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
