Assay Details
Gene Symbol: P4hb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012998.1 | NP_037130.1 | 9 - 10 | 1402 | 97 | |
GenBank mRNA | BC061857.1 | - | 9 - 10 | 1416 | 97 | |
FQ209800.1 | - | 2 - 3 | 216 | 97 | ||
FQ220001.1 | - | 1 - 2 | 189 | 97 | ||
M21018.1 | - | 9 - 10 | 1280 | 97 | ||
X02918.1 | - | 9 - 10 | 1402 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
