Assay Details
Gene Symbol: Ttc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005529.1 | NP_001005529.1 | 3 - 4 | 487 | 88 | |
XM_008767639.1 | XP_008765861.1 | 3 - 4 | 538 | 88 | ||
GenBank mRNA | BC082093.1 | - | 3 - 4 | 487 | 88 | |
FQ213119.1 | - | 3 - 4 | 504 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
