Assay Details
Gene Symbol: Galnt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130742.1 | NP_570098.1 | 11 - 12 | 1648 | 67 | |
GenBank mRNA | AF241241.1 | - | 11 - 12 | 1648 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P144411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79415 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144411 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144411 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P144411 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P550396 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84009 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101452 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91708 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88155 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48448 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41520 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63347 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19366 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40338 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10733736 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733742 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733739 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733731 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733740 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733734 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733735 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733738 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733732 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733733 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733741 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10733730 | - | - |
Rat Genome 230 2.0 Array | 1387290_at | - | - |
Rat Genome 230 2.0 Array | 1387290_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393651_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387290_at | - | - |
Affymetrix HT Rat Focus Array | 1387290_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393651_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000005515.rn.1 | TC1000000645.rn.1 | RTA TC |
Rat Expression Set 230 | 1387290_PM_at | - | - |
Rat Expression Set 230 | 1387290_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10733730 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21705759 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355139 | - | - |
RatRef-12 Expression BeadChip | ILMN_48669 | - | - |
