Assay Details
Gene Symbol: Ube2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022690.2 | NP_073181.1 | 1 - 2 | 347 | 102 | |
GenBank mRNA | BC086980.1 | - | 1 - 2 | 347 | 102 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC086980.1, NM_022690.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
