Assay Details
Gene Symbol: Senp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013116.2 | NP_001013134.2 | 9 - 10 | 1722 | 71 | |
XM_006246703.2 | XP_006246765.1 | 9 - 10 | 1771 | 71 | ||
XM_008767826.1 | XP_008766048.1 | 9 - 10 | 1860 | 71 | ||
GenBank mRNA | BC101901.1 | - | 8 - 9 | 1547 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
