Assay Details
Gene Symbol: Nlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191924.1 | NP_001178853.1 | 10 - 11 | 2718 | 110 | |
XM_006246945.2 | XP_006247007.1 | 10 - 11 | 2520 | 110 | ||
GenBank mRNA | FQ225006.1 | - | 9 - 10 | 1363 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
transforming growth factor beta receptor signaling pathway
Wnt signaling pathway
peptidyl-threonine phosphorylation
negative regulation of Wnt signaling pathway
intracellular signal transduction
serine phosphorylation of STAT protein
protein autophosphorylation
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
ATP binding
transcription factor binding
ubiquitin protein ligase binding
SH2 domain binding
MAPK cascade
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
transforming growth factor beta receptor signaling pathway
Wnt signaling pathway
peptidyl-threonine phosphorylation
negative regulation of Wnt signaling pathway
intracellular signal transduction
serine phosphorylation of STAT protein
protein autophosphorylation
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
ATP binding
transcription factor binding
ubiquitin protein ligase binding
SH2 domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P004927 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004922 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P708147 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004927 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P708147 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004927 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004922 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P004927 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P004922 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P708147 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P429450 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P708147 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P483290 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1050333 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Rat Genome 230 2.0 Array | 1385266_at | - | - |
Rat Genome 230 2.0 Array | 1389926_at | - | - |
Rat Genome 230 2.0 Array | 1382365_at | - | - |
Rat Genome 230 2.0 Array | 1379218_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389926_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379765_at | - | - |
Rat Genome 230 2.0 Array | 1379218_at | - | - |
Rat Genome 230 2.0 Array | 1389926_PM_at | - | - |
Rat Genome 230 2.0 Array | 1385266_at | - | - |
Rat Genome 230 2.0 Array | 1385266_PM_at | - | - |
Rat Genome 230 2.0 Array | 1385266_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389926_at | - | - |
Rat Genome 230 2.0 Array | 1379765_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382365_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379218_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379765_at | - | - |
Affymetrix HT Rat Focus Array | 1379765_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379218_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000029987.rn.1 | TC1000003343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000029991.rn.1 | TC1000003343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000029988.rn.1 | TC1000003343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000029986.rn.1 | TC1000003343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000029989.rn.1 | TC1000003343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000029990.rn.1 | TC1000003343.rn.1 | RTA TC |
Rat Expression Set 230 | 1389926_PM_at | - | - |
Rat Expression Set 230 | 1389926_PM_at | - | - |
Rat Expression Set 230 | 1389926_at | - | - |
Rat Expression Set 230 | 1389926_at | - | - |
Rat Expression Set 230 | 1379765_at | - | - |
Rat Expression Set 230 | 1379218_at | - | - |
Rat Expression Set 230 | 1382365_at | - | - |
Rat Expression Set 230 | 1385266_PM_at | - | - |
Rat Expression Set 230 | 1379218_PM_at | - | - |
Rat Expression Set 230 | 1385266_at | - | - |
Rat Expression Set 230 | 1382365_PM_at | - | - |
Rat Expression Set 230 | 1385266_PM_at | - | - |
Rat Expression Set 230 | 1379765_PM_at | - | - |
Rat Expression Set 230 | 1385266_at | - | - |
Rat Genome U34 Set | rc_AA997745_at | - | - |
Rat Genome U34 Set | rc_AI236404_at | - | - |
Rat Genome U34 Set | rc_AI236404_at | - | - |
Rat Genome U34 Set | rc_AI231394_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22243118 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367449 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353516 | - | - |
RatRef-12 Expression BeadChip | ILMN_48397 | - | - |
RatRef-12 Expression BeadChip | ILMN_58229 | - | - |
