Assay Details
Gene Symbol: Aldh3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031731.2 | NP_113919.2 | 6 - 7 | 1062 | 133 | |
XM_006246529.2 | XP_006246591.1 | 6 - 7 | 1560 | 133 | ||
GenBank mRNA | FQ210485.1 | - | 6 - 7 | 1125 | 133 | |
FQ221576.1 | - | 6 - 7 | 1084 | 133 | ||
M73714.1 | - | 6 - 7 | 1068 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
