Assay Details
Gene Symbol: Tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107012.1 | NP_001100482.1 | 2 - 3 | 282 | 127 | |
XM_006246704.2 | XP_006246766.1 | 3 - 4 | 1740 | 127 | ||
XM_006246705.2 | XP_006246767.1 | 4 - 5 | 815 | 127 | ||
XM_006246707.2 | XP_006246769.1 | 4 - 5 | 485 | 127 | ||
XM_006246709.2 | XP_006246771.1 | 3 - 4 | 662 | 127 | ||
XM_008767827.1 | XP_008766049.1 | 3 - 4 | 1191 | 127 | ||
XM_008767828.1 | XP_008766050.1 | 4 - 5 | 579 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
