Assay Details
Gene Symbol: Slc9a3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245893.2 | XP_006245955.1 | 1 - 2 | 862 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
