Assay Details
Gene Symbol: G6pc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176077.3 | NP_788266.2 | 5 - 6 | 847 | 73 | |
GenBank mRNA | BC078689.1 | - | 5 - 6 | 847 | 73 | |
FQ231610.1 | - | 5 - 6 | 743 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
