Assay Details
Gene Symbol: Senp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013116.2 | NP_001013134.2 | 3 - 4 | 1108 | 132 | |
XM_006246703.2 | XP_006246765.1 | 3 - 4 | 1157 | 132 | ||
XM_008767826.1 | XP_008766048.1 | 3 - 4 | 1246 | 132 | ||
GenBank mRNA | BC101901.1 | - | 2 - 3 | 933 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
