Assay Details
Gene Symbol: Psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134458.2 | NP_604453.2 | 4 - 5 | 361 | 82 | |
XM_006247249.2 | XP_006247311.1 | 4 - 5 | 1213 | 82 | ||
GenBank mRNA | AF352812.1 | - | 4 - 5 | 361 | 82 | |
CK840952.1 | - | 4 - 5 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
