Assay Details
Gene Symbol: Pipox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012009.1 | NP_001012009.1 | 7 - 8 | 1046 | 116 | |
XM_006246958.1 | XP_006247020.1 | 8 - 9 | 1182 | 116 | ||
XM_006246959.1 | XP_006247021.1 | 6 - 7 | 1211 | 116 | ||
GenBank mRNA | BC088249.1 | - | 7 - 8 | 1046 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
