Assay Details
Gene Symbol: Prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013181.1 | NP_037313.1 | 9 - 10 | 987 | 109 | |
XM_008768311.1 | XP_008766533.1 | 9 - 10 | 1043 | 109 | ||
GenBank mRNA | M17086.1 | - | 9 - 10 | 987 | 109 | |
U75932.1 | - | 6 - 7 | 501 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
