Assay Details
Gene Symbol: Jmjd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012143.2 | NP_001012143.2 | 1 - 2 | 418 | 86 | |
XM_006247806.1 | XP_006247868.1 | 1 - 2 | 251 | 86 | ||
GenBank mRNA | BC079012.1 | - | 1 - 2 | 418 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
