Assay Details
Gene Symbol: Jmjd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012143.2 | NP_001012143.2 | 2 - 3 | 807 | 102 | |
XM_006247806.1 | XP_006247868.1 | 2 - 3 | 640 | 102 | ||
GenBank mRNA | BC079012.1 | - | 2 - 3 | 807 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
