Assay Details
Gene Symbol: Nxf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002730283.4 | XP_002730329.2 | 18 - 19 | 1594 | 97 | |
XM_006257244.2 | XP_006257306.1 | 18 - 19 | 1514 | 97 | ||
XM_006257245.2 | XP_006257307.1 | 18 - 19 | 1507 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
