Assay Details
Gene Symbol: Xrcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006999.1 | NP_001007000.1 | 2 - 3 | 174 | 143 | |
XM_006231751.1 | XP_006231813.1 | 2 - 3 | 343 | 143 | ||
GenBank mRNA | BC083848.1 | - | 2 - 3 | 174 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
