Assay Details
Gene Symbol: Xrcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006999.1 | NP_001007000.1 | 6 - 7 | 749 | 108 | |
XM_006231751.1 | XP_006231813.1 | 6 - 7 | 918 | 108 | ||
XM_008760636.1 | XP_008758858.1 | 6 - 7 | 880 | 108 | ||
XM_008760637.1 | XP_008758859.1 | 6 - 7 | 945 | 108 | ||
XM_008760638.1 | XP_008758860.1 | 6 - 7 | 880 | 108 | ||
GenBank mRNA | BC083848.1 | - | 6 - 7 | 749 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
