Assay Details
Gene Symbol: Emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053719.1 | NP_446171.1 | 6 - 7 | 1002 | 132 | |
GenBank mRNA | AJ009698.1 | - | 6 - 7 | 1002 | 132 | |
BC061846.1 | - | 6 - 7 | 1004 | 132 | ||
FQ213962.1 | - | 6 - 7 | 1015 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P304220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171304 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P304220 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P304220 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P304220 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P304220 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35550 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78622 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49172 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10813157 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813159 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813161 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813154 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813160 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821541 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813158 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813156 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813155 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10813152 | - | - |
Rat Genome 230 2.0 Array | 1368541_at | - | - |
Rat Genome 230 2.0 Array | 1368541_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368541_at | - | - |
Affymetrix HT Rat Focus Array | 1368541_PM_at | - | - |
Rat Expression Set 230 | 1368541_at | - | - |
Rat Expression Set 230 | 1368541_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813152 | - | - |
Rat Genome U34 Set | AJ009698_at | - | - |
Rat Genome U34 Set | AJ009698_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21676545 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359464 | - | - |
RatRef-12 Expression BeadChip | ILMN_58897 | - | - |
