Assay Details
Gene Symbol: Rnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031097.1 | NP_112359.1 | 8 - 9 | 1432 | 123 | |
GenBank mRNA | AY724503.1 | - | 8 - 9 | 1607 | 123 | |
BC061718.1 | - | 8 - 9 | 1465 | 123 | ||
D87515.1 | - | 8 - 9 | 1429 | 123 | ||
U61696.1 | - | 8 - 9 | 1432 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
