Assay Details
Gene Symbol: Gpr137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109562.2 | NP_001103032.1 | 5 - 6 | 1071 | 120 | |
GenBank mRNA | BC167037.1 | - | 5 - 6 | 1071 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
