Assay Details
Gene Symbol: Cd163l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106312.1 | NP_001099782.1 | 3 - 4 | 990 | 90 | |
XM_008759975.1 | XP_008758197.1 | 5 - 6 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
